The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000328.2 | NP_000319.1 | 1 - 2 | 195 | 129 |
NM_001034853.1 | NP_001030025.1 | 1 - 2 | 195 | 129 | |
XM_005272633.2 | XP_005272690.1 | 1 - 2 | 224 | 129 | |
XM_011543940.2 | XP_011542242.1 | 1 - 2 | 194 | 129 | |
XM_017029710.1 | XP_016885199.1 | 1 - 2 | 224 | 129 | |
XM_017029711.1 | XP_016885200.1 | 1 - 2 | 224 | 129 | |
XM_017029712.1 | XP_016885201.1 | 1 - 2 | 224 | 129 | |
GenBank mRNA | AJ238395.1 | - | 1 - 2 | 82 | 129 |
AK291832.1 | - | 1 - 2 | 202 | 129 | |
BC031624.1 | - | 1 - 2 | 166 | 129 | |
BP233620.1 | - | 1 - 2 | 204 | 129 | |
BX644678.1 | - | 1 - 2 | 194 | 129 | |
U57629.1 | - | 1 - 2 | 86 | 129 |