Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZSCAN10
Entrez Gene ID
84891
Gene Name
zinc finger and SCAN domain containing 10
Gene Aliases
ZNF206
Chromosome Location
Chr.16: 3088890 - 3099317 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282415.1 NP_001269344.1 3 - 4 459 70
NM_001282416.1 NP_001269345.1 3 - 4 654 70
NM_032805.2 NP_116194.2 4 - 5 922 70
XM_017023791.1 XP_016879280.1 3 - 4 958 70
XM_017023792.1 XP_016879281.1 3 - 4 958 70
GenBank mRNA AK027455.1 - 3 - 4 650 70
AK074736.1 - 3 - 4 434 70
BC111367.1 - 2 - 3 1251 70
BC114452.1 - 2 - 3 361 70
DA729263.1 - 3 - 4 629 70

View Full Product Details