Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAF12
Entrez Gene ID
6883
Gene Name
TATA-box binding protein associated factor 12
Gene Aliases
TAF2J, TAFII20
Chromosome Location
Chr.1: 28603096 - 28648707 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017002186.1 XP_016857675.1 1 - 2 502 116

View Full Product Details