Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IARS
Entrez Gene ID
3376
Gene Name
isoleucyl-tRNA synthetase
Gene Aliases
IARS1, ILERS, ILRS, IRS, PRO0785
Chromosome Location
Chr.9: 92210207 - 92293756 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002161.5 NP_002152.2 18 - 19 2020 90
NM_013417.3 NP_038203.2 18 - 19 2190 90
NR_073446.1 17 - 18 1863 90
GenBank mRNA AB209234.1 - 18 - 19 1929 90
AK293014.1 - 18 - 19 1947 90
BC065552.1 - 17 - 18 1862 90
BX537417.1 - 18 - 19 1958 90
BX537429.1 - 18 - 19 1960 90
BX538109.1 - 17 - 18 1803 90
BX647071.1 - 18 - 19 1959 90
BX647236.1 - 18 - 19 1952 90
BX647430.1 - 18 - 19 1954 90
BX648567.1 - 18 - 19 1960 90
D28473.1 - 18 - 19 2108 90
U04953.1 - 18 - 19 2122 90

View Full Product Details