The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004246.2 | NP_004237.1 | 7 - 8 | 1437 | 63 |
XM_005256861.3 | XP_005256918.1 | 5 - 6 | 613 | 63 | |
XM_011524077.2 | XP_011522379.1 | 8 - 9 | 1551 | 63 | |
XM_017025339.1 | XP_016880828.1 | 7 - 8 | 1494 | 63 | |
XM_017025340.1 | XP_016880829.1 | 5 - 6 | 1270 | 63 | |
XM_017025341.1 | XP_016880830.1 | 4 - 5 | 1163 | 63 | |
GenBank mRNA | AF105367.1 | - | 7 - 8 | 924 | 63 |
AK295765.1 | - | 7 - 8 | 816 | 63 | |
AK314269.1 | - | 7 - 8 | 1437 | 63 | |
BC019818.1 | - | 4 - 5 | 482 | 63 | |
BC041176.1 | - | 4 - 5 | 493 | 63 | |
BC043390.1 | - | 5 - 6 | 632 | 63 | |
BC096261.1 | - | 7 - 8 | 924 | 63 | |
BC096262.1 | - | 7 - 8 | 924 | 63 | |
BC096263.1 | - | 7 - 8 | 920 | 63 | |
BC096264.1 | - | 7 - 8 | 924 | 63 |