Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPK1
Entrez Gene ID
27010
Gene Name
thiamin pyrophosphokinase 1
Gene Aliases
HTPK1, PP20, THMD5
Chromosome Location
Chr.7: 144451941 - 144836424 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042482.1 NP_001035947.1 2 - 3 149 119
NM_022445.3 NP_071890.2 2 - 3 149 119
XM_005249970.1 XP_005250027.1 2 - 3 127 119
XM_011516031.1 XP_011514333.1 2 - 3 150 119
XM_011516032.2 XP_011514334.1 2 - 3 286 119
XM_011516033.2 XP_011514335.1 2 - 3 156 119
XM_011516034.2 XP_011514336.1 2 - 3 213 119
XM_011516035.2 XP_011514337.1 2 - 3 150 119
XM_011516040.2 XP_011514342.1 2 - 3 150 119
XM_011516043.1 XP_011514345.1 2 - 3 150 119
XM_011516046.1 XP_011514348.1 2 - 3 150 119
XM_017011969.1 XP_016867458.1 2 - 3 150 119
XM_017011972.1 XP_016867461.1 2 - 3 150 119
XM_017011973.1 XP_016867462.1 2 - 3 294 119
XM_017011976.1 XP_016867465.1 2 - 3 149 119
XM_017011977.1 XP_016867466.1 2 - 3 121 119
XM_017011979.1 XP_016867468.1 2 - 3 281 119
GenBank mRNA AB028138.1 - 1 - 2 64 119
AF297710.1 - 2 - 3 136 119
AK091736.1 - 2 - 3 88 119
AK289652.1 - 2 - 3 124 119
AK308693.1 - 2 - 3 149 119
AY206415.1 - 2 - 3 131 119
BC068460.1 - 2 - 3 128 119
DA135998.1 - 2 - 3 149 119

View Full Product Details