Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPK1
Entrez Gene ID
27010
Gene Name
thiamin pyrophosphokinase 1
Gene Aliases
HTPK1, PP20, THMD5
Chromosome Location
Chr.7: 144451941 - 144836424 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042482.1 NP_001035947.1 3 - 4 216 79
NM_022445.3 NP_071890.2 3 - 4 216 79
XM_005249970.1 XP_005250027.1 3 - 4 194 79
XM_011516031.1 XP_011514333.1 3 - 4 217 79
XM_011516032.2 XP_011514334.1 3 - 4 353 79
XM_011516033.2 XP_011514335.1 3 - 4 223 79
XM_011516034.2 XP_011514336.1 3 - 4 280 79
XM_011516035.2 XP_011514337.1 3 - 4 217 79
XM_011516040.2 XP_011514342.1 3 - 4 217 79
XM_011516043.1 XP_011514345.1 3 - 4 217 79
XM_011516046.1 XP_011514348.1 3 - 4 217 79
XM_017011969.1 XP_016867458.1 3 - 4 217 79
XM_017011970.1 XP_016867459.1 1 - 2 70 79
XM_017011972.1 XP_016867461.1 3 - 4 217 79
XM_017011973.1 XP_016867462.1 3 - 4 361 79
XM_017011974.1 XP_016867463.1 1 - 2 70 79
XM_017011976.1 XP_016867465.1 3 - 4 216 79
XM_017011977.1 XP_016867466.1 3 - 4 188 79
XM_017011978.1 XP_016867467.1 2 - 3 147 79
XM_017011979.1 XP_016867468.1 3 - 4 348 79
XM_017011985.1 XP_016867474.1 2 - 3 147 79
GenBank mRNA AB028138.1 - 2 - 3 131 79
AF297710.1 - 3 - 4 203 79
AK091736.1 - 3 - 4 155 79
AK289652.1 - 3 - 4 191 79
AK308693.1 - 3 - 4 216 79
AY206415.1 - 3 - 4 198 79
BC068460.1 - 3 - 4 195 79
BI770816.1 - 2 - 3 132 79
BX446289.2 - 3 - 4 198 79
DA135998.1 - 3 - 4 216 79

View Full Product Details