Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AQP6
Entrez Gene ID
363
Gene Name
aquaporin 6
Gene Aliases
AQP2L, KID
Chromosome Location
Chr.12: 49972837 - 49977139 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AL137716.1 - 4 - 5 2414 84

View Full Product Details