The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244134.1 | NP_001231063.1 | 7 - 8 | 1657 | 72 |
NM_001320961.1 | NP_001307890.1 | 7 - 8 | 1413 | 72 | |
NM_005204.3 | NP_005195.2 | 8 - 9 | 1888 | 72 | |
XM_017015707.1 | XP_016871196.1 | 7 - 8 | 2105 | 72 | |
XM_017015708.1 | XP_016871197.1 | 8 - 9 | 1618 | 72 | |
XM_017015709.1 | XP_016871198.1 | 7 - 8 | 4398 | 72 | |
XM_017015710.1 | XP_016871199.1 | 7 - 8 | 1508 | 72 | |
XM_017015711.1 | XP_016871200.1 | 7 - 8 | 5467 | 72 | |
XM_017015712.1 | XP_016871201.1 | 8 - 9 | 4980 | 72 | |
XM_017015713.1 | XP_016871202.1 | 8 - 9 | 5224 | 72 | |
XM_017015714.1 | XP_016871203.1 | 7 - 8 | 5019 | 72 | |
GenBank mRNA | AK290320.1 | - | 7 - 8 | 1553 | 72 |
AK313295.1 | - | 7 - 8 | 1554 | 72 | |
BC104833.1 | - | 7 - 8 | 1514 | 72 | |
BC113566.1 | - | 7 - 8 | 1514 | 72 | |
CR542284.1 | - | 6 - 7 | 1276 | 72 | |
D14497.1 | - | 8 - 9 | 1642 | 72 | |
Z14138.1 | - | 6 - 7 | 1436 | 72 |