Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP3K8
Entrez Gene ID
1326
Gene Name
mitogen-activated protein kinase kinase kinase 8
Gene Aliases
AURA2, COT, EST, ESTF, MEKK8, TPL2, Tpl-2, c-COT
Chromosome Location
Chr.10: 30434021 - 30461833 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320961.1 NP_001307890.1 1 - 2 109 69
GenBank mRNA BI753824.1 - 1 - 2 79 69

View Full Product Details