The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007167.3 | NP_009098.3 | 12 - 13 | 2033 | 93 |
GenBank mRNA | AK299293.1 | - | 12 - 13 | 1992 | 93 |
AK301461.1 | - | 11 - 12 | 1801 | 93 | |
AK310703.1 | - | 12 - 13 | 1974 | 93 | |
AK310938.1 | - | 10 - 11 | 1533 | 93 | |
AL833263.1 | - | 11 - 12 | 1756 | 93 | |
BC033903.1 | - | 12 - 13 | 2015 | 93 | |
BC107880.1 | - | 12 - 13 | 2014 | 93 |