Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYT17
Entrez Gene ID
51760
Gene Name
synaptotagmin 17
Gene Aliases
sytXVII
Chromosome Location
Chr.16: 19167834 - 19268334 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016524.3 NP_057608.2 1 - 2 452 76
XM_017023280.1 XP_016878769.1 1 - 2 446 76
GenBank mRNA CA389452.1 - 1 - 2 349 76

View Full Product Details