Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMP20
Entrez Gene ID
9313
Gene Name
matrix metallopeptidase 20
Gene Aliases
AI2A2, MMP-20
Chromosome Location
Chr.11: 102576835 - 102625332 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004771.3 NP_004762.2 7 - 8 1108 93
GenBank mRNA Y12779.1 - 7 - 8 1108 93

View Full Product Details