The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005588.2 | NP_005579.2 | 7 - 8 | 593 | 111 |
XM_011514628.1 | XP_011512930.1 | 6 - 7 | 650 | 111 | |
XM_011514629.1 | XP_011512931.1 | 7 - 8 | 593 | 111 | |
GenBank mRNA | AF478685.1 | - | 7 - 8 | 561 | 111 |
AK225212.1 | - | 7 - 8 | 583 | 111 | |
AK290282.1 | - | 7 - 8 | 561 | 111 | |
AK315246.1 | - | 7 - 8 | 565 | 111 | |
AK315840.1 | - | 7 - 8 | 563 | 111 | |
BC131714.1 | - | 7 - 8 | 564 | 111 | |
BC143650.1 | - | 5 - 6 | 732 | 111 | |
BC143651.1 | - | 6 - 7 | 643 | 111 | |
M82962.1 | - | 7 - 8 | 560 | 111 |