Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEP1A
Entrez Gene ID
4224
Gene Name
meprin A subunit alpha
Gene Aliases
PPHA
Chromosome Location
Chr.6: 46793357 - 46845987 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005588.2 NP_005579.2 8 - 9 827 70
XM_011514628.1 XP_011512930.1 7 - 8 884 70
XM_011514629.1 XP_011512931.1 8 - 9 827 70
GenBank mRNA AF478685.1 - 8 - 9 795 70
AK225212.1 - 8 - 9 817 70
AK290282.1 - 8 - 9 795 70
AK315246.1 - 8 - 9 799 70
AK315840.1 - 8 - 9 797 70
BC131714.1 - 8 - 9 798 70
BC143650.1 - 6 - 7 966 70
BC143651.1 - 7 - 8 877 70
M82962.1 - 8 - 9 794 70

View Full Product Details