The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199692.1 | NP_001186621.1 | 16 - 17 | 3576 | 93 |
NM_001199693.1 | NP_001186622.1 | 15 - 16 | 2602 | 93 | |
NM_001199694.1 | NP_001186623.1 | 15 - 16 | 3023 | 93 | |
NM_003040.3 | NP_003031.3 | 16 - 17 | 2792 | 93 | |
GenBank mRNA | AB209158.1 | - | 16 - 17 | 3576 | 93 |
AK295767.1 | - | 15 - 16 | 2602 | 93 | |
AK312699.1 | - | 16 - 17 | 2771 | 93 | |
BC004893.2 | - | 16 - 17 | 2429 | 93 | |
BC009386.2 | - | 15 - 16 | 3023 | 93 | |
BC009434.1 | - | 15 - 16 | 3023 | 93 | |
BC010069.2 | - | 16 - 17 | 2429 | 93 | |
BC028601.2 | - | 15 - 16 | 2624 | 93 | |
U62531.1 | - | 16 - 17 | 2710 | 93 | |
X03918.1 | - | 9 - 10 | 1408 | 93 | |
X62137.1 | - | 16 - 17 | 2612 | 93 |