Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARSD
Entrez Gene ID
414
Gene Name
arylsulfatase D
Gene Aliases
ASD
Chromosome Location
Chr.X: 2903970 - 2929375 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC080179.1 - 3 - 4 321 174

View Full Product Details