The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001185009.2 | NP_001171938.1 | 4 - 5 | 993 | 136 |
NM_001185010.2 | NP_001171939.1 | 3 - 4 | 471 | 136 | |
NM_002938.4 | NP_002929.1 | 3 - 4 | 471 | 136 | |
GenBank mRNA | AB000468.1 | - | 3 - 4 | 420 | 136 |
AK128038.1 | - | 3 - 4 | 1634 | 136 | |
AK307372.1 | - | 3 - 4 | 433 | 136 | |
AK309509.1 | - | 3 - 4 | 399 | 136 | |
AK312534.1 | - | 3 - 4 | 400 | 136 | |
BC031935.1 | - | 4 - 5 | 993 | 136 | |
DA342512.1 | - | 3 - 4 | 471 | 136 | |
U95140.1 | - | 3 - 4 | 393 | 136 |