Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC25A38
Entrez Gene ID
54977
Gene Name
solute carrier family 25 member 38
Gene Aliases
SIDBA2
Chromosome Location
Chr.3: 39383324 - 39397328 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006713214.1 XP_006713277.1 2 - 3 436 130

View Full Product Details