Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CASP8AP2
Entrez Gene ID
9994
Gene Name
caspase 8 associated protein 2
Gene Aliases
CED-4, FLASH, RIP25
Chromosome Location
Chr.6: 89829900 - 89874436 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001137667.1 NP_001131139.1 4 - 5 422 134
NM_001137668.1 NP_001131140.1 4 - 5 289 134
NM_012115.3 NP_036247.1 4 - 5 461 134
GenBank mRNA AB037736.2 - 3 - 4 226 134
AF154415.1 - 4 - 5 422 134
AF164678.2 - 3 - 4 226 134
BC056685.1 - 4 - 5 295 134
BC132828.1 - 3 - 4 269 134
BC132830.1 - 4 - 5 308 134
BC150274.1 - 3 - 4 255 134
BP192859.1 - 4 - 5 289 134
BP363682.1 - 4 - 5 422 134

View Full Product Details