The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003559.4 | NP_003550.1 | 6 - 7 | 1175 | 141 |
XM_011525326.2 | XP_011523628.1 | 7 - 8 | 1316 | 141 | |
XM_011525327.2 | XP_011523629.1 | 8 - 9 | 835 | 141 | |
XM_011525329.2 | XP_011523631.1 | 7 - 8 | 1316 | 141 | |
XM_011525330.2 | XP_011523632.1 | 5 - 6 | 511 | 141 | |
XM_017025196.1 | XP_016880685.1 | 7 - 8 | 752 | 141 | |
XM_017025197.1 | XP_016880686.1 | 7 - 8 | 652 | 141 | |
XM_017025198.1 | XP_016880687.1 | 5 - 6 | 1138 | 141 | |
XM_017025199.1 | XP_016880688.1 | 6 - 7 | 590 | 141 | |
GenBank mRNA | BC027459.1 | - | 6 - 7 | 792 | 141 |
BT019614.1 | - | 6 - 7 | 694 | 141 | |
U85245.1 | - | 6 - 7 | 1174 | 141 |