The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207118.2 | NP_997001.1 | 2 - 3 | 119 | 138 |
GenBank mRNA | AJ634743.1 | - | 2 - 3 | 95 | 138 |
AK055106.1 | - | 2 - 3 | 114 | 138 | |
BC056906.1 | - | 2 - 3 | 106 | 138 | |
BC060317.1 | - | 1 - 2 | 71 | 138 | |
CN403888.1 | - | 2 - 3 | 122 | 138 |