The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281444.1 | NP_001268373.1 | 1 - 2 | 198 | 145 |
NM_181804.2 | NP_861520.1 | 1 - 2 | 198 | 145 | |
NM_181805.2 | NP_861521.1 | 1 - 2 | 198 | 145 | |
GenBank mRNA | BU161358.1 | - | 1 - 2 | 144 | 145 |