Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUPT6H
Entrez Gene ID
6830
Gene Name
SPT6 homolog, histone chaperone
Gene Aliases
SPT6, SPT6H, emb-5
Chromosome Location
Chr.17: 28662183 - 28702684 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320755.1 NP_001307684.1 11 - 12 1490 129
NM_003170.4 NP_003161.2 10 - 11 1388 129
XM_017024978.1 XP_016880467.1 12 - 13 1549 129
XM_017024979.1 XP_016880468.1 10 - 11 1626 129
XM_017024980.1 XP_016880469.1 9 - 10 1320 129
GenBank mRNA AL833607.1 - 11 - 12 1491 129
BC033074.1 - 7 - 8 916 129
BC136522.1 - 10 - 11 1300 129
BC136524.1 - 10 - 11 1300 129
BC150268.1 - 10 - 11 1302 129
D79984.1 - 10 - 11 1302 129

View Full Product Details