The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272046.1 | NP_001258975.1 | 9 - 10 | 1217 | 77 |
NM_001320804.1 | NP_001307733.1 | 9 - 10 | 1159 | 77 | |
XM_011539754.2 | XP_011538056.1 | 10 - 11 | 1394 | 77 | |
XM_011539757.2 | XP_011538059.1 | 9 - 10 | 1489 | 77 | |
XM_017016177.1 | XP_016871666.1 | 9 - 10 | 969 | 77 | |
XM_017016178.1 | XP_016871667.1 | 9 - 10 | 969 | 77 | |
XM_017016179.1 | XP_016871668.1 | 9 - 10 | 922 | 77 | |
XM_017016180.1 | XP_016871669.1 | 1 - 2 | 61 | 77 | |
XM_017016181.1 | XP_016871670.1 | 9 - 10 | 966 | 77 | |
GenBank mRNA | AJ536328.2 | - | 9 - 10 | 1141 | 77 |
AK122716.1 | - | 9 - 10 | 1212 | 77 | |
AK127756.1 | - | 9 - 10 | 1048 | 77 | |
AY572972.1 | - | 9 - 10 | 979 | 77 | |
BC128588.1 | - | 8 - 9 | 981 | 77 | |
BC143380.1 | - | 9 - 10 | 919 | 77 |