The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207352.3 | NP_997235.3 | 8 - 9 | 1396 | 153 |
XM_005262935.3 | XP_005262992.1 | 8 - 9 | 1181 | 153 | |
XM_017008037.1 | XP_016863526.1 | 7 - 8 | 1154 | 153 | |
GenBank mRNA | AK022114.1 | - | 2 - 3 | 269 | 153 |
AK122600.1 | - | 8 - 9 | 1131 | 153 | |
AK126473.1 | - | 9 - 10 | 1065 | 153 | |
AK311390.1 | - | 2 - 3 | 182 | 153 | |
AY422002.1 | - | 8 - 9 | 1396 | 153 | |
BC041839.1 | - | 1 - 2 | 830 | 153 | |
BC060857.1 | - | 8 - 9 | 1262 | 153 | |
BX648537.1 | - | 2 - 3 | 327 | 153 | |
BX648730.1 | - | 3 - 4 | 5150 | 153 |