Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AKR7A2P1
- Entrez Gene ID
-
246182
- Gene Name
- aldo-keto reductase family 7 member A2 pseudogene 1
- Gene Aliases
-
AFARP1
- Chromosome Location
-
Chr.1: 112923350 - 112924673 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002796.2 |
|
1 - 1 |
274 |
153 |
GenBank mRNA |
AJ277796.1 |
- |
1 - 1 |
274 |
153 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BF765305.1 |
- |
1 |
116 |
|
Chr. 1 - 112923517 - 112923786 [+] on Build GRCh38 |
153 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized HP08777 |
HP08777 |
|
-45.5kb
|
Sense |
Chr. - 112850344 - 112877871 on Build GRCh38 |
Downstream |
SLC16A1 antisense RNA 1 |
SLC16A1-AS1 |
|
31.7kb
|
Sense |
Chr. - 112956415 - 112964072 on Build GRCh38 |
View Full Product Details