The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152316.2 | NP_689529.1 | 4 - 4 | 1712 | 89 |
XM_005252792.3 | XP_005252849.1 | 4 - 4 | 2163 | 89 | |
GenBank mRNA | AK096287.1 | - | 4 - 4 | 1709 | 89 |
BX647617.1 | - | 4 - 4 | 1706 | 89 |