Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PCDH11Y
- Entrez Gene ID
-
83259
- Gene Name
- protocadherin 11 Y-linked
- Gene Aliases
-
PCDH-PC, PCDH22, PCDHX, PCDHY
- Chromosome Location
-
Chr.Y: 5000044 - 5742228 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AJ564961.1 |
- |
1 - 1 |
2957 |
124 |
- Gene Symbol
- PCDH11X
- Entrez Gene ID
-
27328
- Gene Name
- protocadherin 11 X-linked
- Gene Aliases
-
PCDH-X, PCDH11, PCDHX, PPP1R119
- Chromosome Location
-
Chr.X: 91779310 - 92623230 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB026187.1 |
- |
2 - 2 |
|
124 |
AJ564934.1 |
- |
1 - 1 |
|
124 |
View Full Product Details