Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR14J1
Entrez Gene ID
442191
Gene Name
olfactory receptor family 14 subfamily J member 1
Gene Aliases
OR5U1, OR6-25, bA150A6.4, hs6M1-28
Chromosome Location
Chr.6: 29306690 - 29307655 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030946.1 NP_112208.1 1 - 1 845 96
GenBank mRNA BC136905.1 - 1 - 1 909 96

View Full Product Details