The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005636.3 | NP_005627.1 | 2 - 3 | 123 | 119 |
NM_175729.1 | NP_783856.1 | 2 - 3 | 123 | 119 | |
XM_017029759.1 | XP_016885248.1 | 2 - 3 | 181 | 119 | |
GenBank mRNA | AX740218.1 | - | 1 - 2 | 65 | 119 |
BC005325.1 | - | 2 - 3 | 123 | 119 | |
BC137394.1 | - | 1 - 2 | 90 | 119 | |
BC137395.1 | - | 1 - 2 | 90 | 119 | |
U90841.1 | - | 1 - 2 | 65 | 119 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034832.3 | NP_001030004.1 | 2 - 3 | 134 | 119 |
NM_001040612.2 | NP_001035702.1 | 2 - 3 | 134 | 119 | |
XM_017029613.1 | XP_016885102.1 | 2 - 3 | 181 | 119 | |
GenBank mRNA | AA312651.1 | - | 2 - 3 | 228 | 119 |
BE866727.1 | - | 2 - 3 | 104 | 119 |