The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001021.4 | NP_001012.1 | 3 - 4 | 381 | 100 |
NR_111943.1 | 2 - 3 | 587 | 100 | ||
NR_111944.1 | 3 - 4 | 381 | 100 | ||
GenBank mRNA | AK311951.1 | - | 3 - 4 | 289 | 100 |
BC009407.2 | - | 1 - 1 | 272 | 100 | |
BC019899.1 | - | 1 - 1 | 277 | 100 | |
BC020453.1 | - | 1 - 1 | 276 | 100 | |
BC022370.1 | - | 3 - 4 | 294 | 100 | |
BC049824.1 | - | 1 - 1 | 269 | 100 | |
BC062715.1 | - | 3 - 4 | 379 | 100 | |
BC071928.1 | - | 1 - 1 | 280 | 100 | |
BE905890.1 | - | 2 - 3 | 257 | 100 | |
BI597659.1 | - | 2 - 3 | 587 | 100 | |
M13932.1 | - | 3 - 4 | 290 | 100 |