Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCB9
Entrez Gene ID
23457
Gene Name
ATP binding cassette subfamily B member 9
Gene Aliases
EST122234, TAPL
Chromosome Location
Chr.12: 122918708 - 122975160 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243013.1 NP_001229942.1 11 - 11 2806 65
NM_019624.3 NP_062570.1 11 - 11 2866 65
NM_019625.3 NP_062571.1 12 - 12 2995 65
XM_005253558.2 XP_005253615.1 12 - 12 2935 65
XM_011538095.2 XP_011536397.1 12 - 12 3218 65
XM_011538096.2 XP_011536398.1 12 - 12 3014 65
XM_011538098.2 XP_011536400.1 11 - 11 3860 65
XM_017019103.1 XP_016874592.1 13 - 13 3056 65
XM_017019104.1 XP_016874593.1 10 - 10 2354 65
GenBank mRNA AB040953.2 - 12 - 12 5524 65
AB177852.1 - 12 - 12 2907 65
AF216494.1 - 12 - 12 2983 65
AK074738.1 - 9 - 9 1939 65
BC064384.1 - 12 - 12 3272 65
CA503047.1 - 1 - 1 65

View Full Product Details