The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004514.3 | NP_004505.2 | 8 - 8 | 1833 | 64 |
GenBank mRNA | U58196.1 | - | 8 - 8 | 1844 | 64 |
U58197.1 | - | 8 - 8 | 1844 | 64 |