Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKD1L2
Entrez Gene ID
114780
Gene Name
polycystin 1 like 2 (gene/pseudogene)
Gene Aliases
PC1L2
Chromosome Location
Chr.16: 81100879 - 81220370 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076780.1 NP_001070248.1 10 - 11 1789 65
NM_052892.3 NP_443124.3 10 - 11 1789 65
NR_126532.1 10 - 11 1789 65
GenBank mRNA AB067466.1 - 2 - 3 134 65
AK098052.1 - 4 - 5 371 65
AY164483.1 - 10 - 11 1789 65

View Full Product Details