The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304548.1 | NP_001291477.1 | 61 - 62 | 9086 | 154 |
GenBank mRNA | BG190566.1 | - | 1 - 2 | 193 | 154 |
BG197183.1 | - | 1 - 2 | 174 | 154 |