Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SNORD15B
- Entrez Gene ID
-
114599
- Gene Name
- small nucleolar RNA, C/D box 15B
- Gene Aliases
-
RNU15B, U15B
- Chromosome Location
-
Chr.11: 75404421 - 75404566 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_000025.1 |
|
1 - 1 |
72 |
67 |
GenBank mRNA |
L15557.1 |
- |
1 - 1 |
72 |
67 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA636472.1 |
- |
|
68 |
|
Chr. - - [-] on Build |
67 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ribosomal protein S3 |
RPS3 |
S3 |
|
Sense |
Chr. - 75399491 - 75422302 on Build GRCh38 |
Downstream |
kelch like family member 35 |
KLHL35 |
|
17.8kb
|
Anti-sense |
Chr. - 75422394 - 75434550 on Build GRCh38 |
View Full Product Details