Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PHF2P1
- Entrez Gene ID
-
266695
- Gene Name
- PHD finger protein 2 pseudogene 1
- Gene Aliases
-
DKFZp686A1627
- Chromosome Location
-
Chr.13: 19045351 - 19072391 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AL832753.1 |
- |
6 - 7 |
846 |
68 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 442 |
LINC00442 |
|
-43.8kb
|
Anti-sense |
Chr. - 19008259 - 19012634 on Build GRCh38 |
Downstream |
tubulin alpha 3c |
TUBA3C |
TUBA2, bA408E5.3 |
111.1kb
|
Sense |
Chr. - 19173770 - 19181852 on Build GRCh38 |
View Full Product Details