The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_036497.1 | 1 - 1 | 2781 | 87 | |
GenBank mRNA | BC042414.1 | - | 1 - 1 | 2752 | 87 |
JX046915.1 | - | 1 - 1 | 392 | 87 |