Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ZMYM6
- Entrez Gene ID
-
9204
- Gene Name
- zinc finger MYM-type containing 6
- Gene Aliases
-
Buster2, MYM, ZBED7, ZNF198L4, ZNF258
- Chromosome Location
-
Chr.1: 34986165 - 35031968 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC043174.1 |
- |
1 - 1 |
449 |
84 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BI597615.1 |
- |
1 |
449 |
|
Chr. 1 - 35006032 - 35006834 [+] on Build GRCh38 |
84 |
GenBankmRNA |
BG715199.1 |
- |
1 |
449 |
|
Chr. 1 - 35006032 - 35006716 [+] on Build GRCh38 |
84 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
transmembrane protein 35B |
TMEM35B |
ZMYM6NB |
-20.7kb
|
Anti-sense |
Chr. - 34981526 - 34985347 on Build GRCh38 |
Downstream |
zinc finger MYM-type containing 1 |
ZMYM1 |
MYM |
52.2kb
|
Sense |
Chr. - 35059786 - 35116458 on Build GRCh38 |
View Full Product Details