Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MORN1
- Entrez Gene ID
-
79906
- Gene Name
- MORN repeat containing 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 2321253 - 2391751 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_125361.1 |
|
7 - 7 |
4824 |
80 |
GenBank mRNA |
AI188092.1 |
- |
1 - 1 |
|
80 |
AK128431.1 |
- |
1 - 1 |
3854 |
80 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB346868.1 |
- |
1 |
|
|
Chr. 1 - 2374856 - 2375379 [+] on Build GRCh38 |
80 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
small nuclear ribonucleoprotein polypeptide N pseudogene |
LOC100129534 |
|
-22.2kb
|
Sense |
Chr. - 2350414 - 2352661 on Build GRCh38 |
Downstream |
retention in endoplasmic reticulum sorting receptor 1 |
RER1 |
|
12.5kb
|
Anti-sense |
Chr. - 2391314 - 2405444 on Build GRCh38 |
View Full Product Details