Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NREP
- Entrez Gene ID
-
9315
- Gene Name
- neuronal regeneration related protein
- Gene Aliases
-
C5orf13, D4S114, P311, PRO1873, PTZ17, SEZ17
- Chromosome Location
-
Chr.5: 111729303 - 111976931 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK021624.1 |
- |
- |
|
69 |
- Gene Symbol
- STARD4-AS1
- Entrez Gene ID
-
100505678
- Gene Name
- STARD4 antisense RNA 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 111512226 - 111739726 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_040093.1 |
|
- |
|
69 |
View Full Product Details