Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CRYM-AS1
- Entrez Gene ID
-
400508
- Gene Name
- CRYM antisense RNA 1
- Gene Aliases
-
NCRNA00169
- Chromosome Location
-
Chr.16: 21300849 - 21318591 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026675.1 |
|
1 - 2 |
493 |
75 |
GenBank mRNA |
AK123760.1 |
- |
1 - 2 |
494 |
75 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA608747.1 |
- |
1 - 2 |
494 |
|
Chr. 16 - 21300849 - 21315951 [+] on Build GRCh38 |
75 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ankyrin repeat and sterile alpha motif domain containing 4B |
ANKS4B |
HARP |
-48.4kb
|
Sense |
Chr. - 21233695 - 21252429 on Build GRCh38 |
Downstream |
sorting nexin 29 pseudogene 1 |
SNX29P1 |
RUNDC2B, RUNDC2L |
33.4kb
|
Sense |
Chr. - 21349363 - 21385933 on Build GRCh38 |
View Full Product Details