The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014834.4 | NP_055649.4 | 4 - 5 | 2831 | 92 |
XM_005257879.4 | XP_005257936.1 | 4 - 5 | 3089 | 92 | |
XM_005257880.4 | XP_005257937.1 | 4 - 5 | 3089 | 92 | |
XM_005257882.4 | XP_005257939.1 | 5 - 6 | 3275 | 92 | |
XM_005257883.4 | XP_005257940.1 | 4 - 5 | 3089 | 92 | |
XM_005257884.4 | XP_005257941.1 | 4 - 5 | 3089 | 92 | |
XM_017025472.1 | XP_016880961.1 | 6 - 7 | 2688 | 92 | |
XM_017025473.1 | XP_016880962.1 | 4 - 5 | 335 | 92 | |
GenBank mRNA | BC040501.1 | - | 4 - 5 | 2831 | 92 |
CR749232.1 | - | 4 - 5 | 2832 | 92 | |
JQ790562.1 | - | 4 - 5 | 388 | 92 | |
JQ790566.1 | - | 4 - 5 | 388 | 92 | |
JQ790572.1 | - | 4 - 5 | 388 | 92 | |
JQ790577.1 | - | 4 - 5 | 388 | 92 | |
JQ790578.1 | - | 4 - 5 | 388 | 92 | |
JQ790579.1 | - | 4 - 5 | 388 | 92 | |
JQ790580.1 | - | 4 - 5 | 388 | 92 | |
JQ790581.1 | - | 4 - 5 | 388 | 92 | |
JQ790688.1 | - | 4 - 5 | 388 | 92 | |
JQ790690.1 | - | 4 - 5 | 388 | 92 | |
JQ790691.1 | - | 4 - 5 | 388 | 92 | |
JQ790692.1 | - | 4 - 5 | 388 | 92 | |
JQ790693.1 | - | 4 - 5 | 388 | 92 | |
JQ790694.1 | - | 4 - 5 | 388 | 92 | |
JQ790699.1 | - | 4 - 5 | 388 | 92 | |
JQ790726.1 | - | 4 - 5 | 390 | 92 | |
JQ790727.1 | - | 4 - 5 | 390 | 92 | |
JQ790729.1 | - | 4 - 5 | 390 | 92 | |
JQ790730.1 | - | 4 - 5 | 390 | 92 | |
JQ790738.1 | - | 4 - 5 | 390 | 92 | |
JQ790741.1 | - | 4 - 5 | 390 | 92 | |
JQ790743.1 | - | 4 - 5 | 390 | 92 | |
JQ790744.1 | - | 4 - 5 | 389 | 92 | |
JQ790749.1 | - | 4 - 5 | 390 | 92 | |
JQ790755.1 | - | 4 - 5 | 390 | 92 | |
JQ790756.1 | - | 4 - 5 | 390 | 92 | |
JQ790757.1 | - | 4 - 5 | 390 | 92 | |
JQ790758.1 | - | 4 - 5 | 390 | 92 | |
JQ790763.1 | - | 4 - 5 | 391 | 92 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006607.2 | NP_001006608.2 | 4 - 5 | 2831 | 92 |
XM_011524842.2 | XP_011523144.1 | 4 - 5 | 2884 | 92 | |
XM_011524843.2 | XP_011523145.1 | 4 - 5 | 2884 | 92 | |
XM_011524849.1 | XP_011523151.1 | 4 - 5 | 2884 | 92 | |
XM_011524850.2 | XP_011523152.1 | 4 - 5 | 2884 | 92 | |
GenBank mRNA | AK126994.1 | - | 4 - 5 | 1513 | 92 |
AY386262.1 | - | 4 - 5 | 1252 | 92 | |
BC144427.1 | - | 4 - 5 | 494 | 92 | |
BC144428.1 | - | 4 - 5 | 494 | 92 | |
JQ790563.1 | - | 4 - 5 | 388 | 92 | |
JQ790564.1 | - | 4 - 5 | 388 | 92 | |
JQ790568.1 | - | 4 - 5 | 388 | 92 | |
JQ790570.1 | - | 4 - 5 | 388 | 92 | |
JQ790571.1 | - | 4 - 5 | 388 | 92 | |
JQ790576.1 | - | 4 - 5 | 388 | 92 | |
JQ790584.1 | - | 4 - 5 | 388 | 92 | |
JQ790598.1 | - | 3 - 4 | 316 | 92 | |
JQ790606.1 | - | 4 - 5 | 388 | 92 | |
JQ790610.1 | - | 4 - 5 | 388 | 92 | |
JQ790617.1 | - | 4 - 5 | 388 | 92 | |
JQ790626.1 | - | 4 - 5 | 388 | 92 | |
JQ790628.1 | - | 4 - 5 | 388 | 92 | |
JQ790700.1 | - | 4 - 5 | 388 | 92 | |
JQ790703.1 | - | 4 - 5 | 364 | 92 | |
JQ790735.1 | - | 4 - 5 | 390 | 92 | |
JQ790739.1 | - | 4 - 5 | 390 | 92 | |
JQ790740.1 | - | 4 - 5 | 390 | 92 | |
JQ790748.1 | - | 4 - 5 | 391 | 92 | |
JQ790750.1 | - | 4 - 5 | 390 | 92 | |
JQ790764.1 | - | 4 - 5 | 390 | 92 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199340.3 | NP_955372.2 | 7 - 8 | 3494 | 92 |
XM_011524776.1 | XP_011523078.1 | 4 - 5 | 933 | 92 | |
XM_011524778.2 | XP_011523080.2 | 5 - 6 | 458 | 92 | |
XM_017024593.1 | XP_016880082.1 | 5 - 6 | 3323 | 92 | |
XM_017024599.1 | XP_016880088.1 | 6 - 7 | 2695 | 92 | |
XM_017024600.1 | XP_016880089.1 | 5 - 6 | 3323 | 92 | |
XM_017024601.1 | XP_016880090.1 | 4 - 5 | 635 | 92 | |
XM_017024603.1 | XP_016880092.1 | 4 - 5 | 933 | 92 | |
XM_017024607.1 | XP_016880096.1 | 4 - 5 | 238 | 92 | |
XM_017024608.1 | XP_016880097.1 | 4 - 5 | 1255 | 92 | |
GenBank mRNA | AB011135.2 | - | 6 - 7 | 3359 | 92 |
AK315945.1 | - | 4 - 5 | 966 | 92 | |
BC043145.1 | - | 4 - 5 | 2836 | 92 | |
BI560889.1 | - | 4 - 5 | 275 | 92 | |
JQ790603.1 | - | 4 - 5 | 388 | 92 | |
JQ790607.1 | - | 4 - 5 | 388 | 92 | |
JQ790608.1 | - | 4 - 5 | 388 | 92 | |
JQ790612.1 | - | 4 - 5 | 388 | 92 | |
JQ790613.1 | - | 4 - 5 | 388 | 92 | |
JQ790704.1 | - | 4 - 5 | 388 | 92 | |
JQ790705.1 | - | 4 - 5 | 388 | 92 | |
JQ790706.1 | - | 4 - 5 | 388 | 92 | |
JQ790713.1 | - | 3 - 4 | 316 | 92 | |
JQ790728.1 | - | 4 - 5 | 390 | 92 | |
JQ790731.1 | - | 4 - 5 | 390 | 92 | |
JQ790734.1 | - | 4 - 5 | 398 | 92 | |
JQ790737.1 | - | 4 - 5 | 390 | 92 | |
JQ790747.1 | - | 4 - 5 | 390 | 92 | |
JQ790751.1 | - | 4 - 5 | 390 | 92 | |
JQ790759.1 | - | 4 - 5 | 390 | 92 | |
JQ790760.1 | - | 4 - 5 | 388 | 92 | |
JQ790761.1 | - | 4 - 5 | 390 | 92 | |
JQ790762.1 | - | 4 - 5 | 391 | 92 | |
JQ790765.1 | - | 4 - 5 | 390 | 92 | |
JQ790766.1 | - | 4 - 5 | 390 | 92 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | JQ790585.1 | - | 4 - 5 | 388 | 92 |
JQ790586.1 | - | 4 - 5 | 388 | 92 | |
JQ790630.1 | - | 4 - 5 | 388 | 92 | |
JQ790698.1 | - | 4 - 5 | 388 | 92 |