Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TMC7
- Entrez Gene ID
-
79905
- Gene Name
- transmembrane channel like 7
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 18983934 - 19063942 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF088010.1 |
- |
1 - 1 |
655 |
88 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
N46353.1 |
- |
1 |
|
|
Chr. 16 - 19066251 - 19066694 [-] on Build GRCh38 |
88 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC102723385 |
LOC102723385 |
|
|
Anti-sense |
Chr. - 19062754 - 19067691 on Build GRCh38 |
Downstream |
coenzyme Q7, hydroxylase |
COQ7 |
CAT5, CLK-1, CLK1, COQ10D8 |
897.0
|
Sense |
Chr. - 19067595 - 19083097 on Build GRCh38 |
View Full Product Details