Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AF038184.1 - 1 452 Chr. 11 - 47529070 - 47530388 [-] on Build GRCh38 84


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream CUGBP, Elav-like family member 1 CELF1 BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50 Sense Chr. - 47465937 - 47565569 on Build GRCh38
Downstream protein tyrosine phosphatase, mitochondrial 1 PTPMT1 DUSP23, MOSP, PLIP, PNAS-129 35.0kb Anti-sense Chr. - 47565430 - 47573461 on Build GRCh38

View Full Product Details