Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPATCH2L
Entrez Gene ID
55668
Gene Name
G-patch domain containing 2 like
Gene Aliases
C14orf118
Chromosome Location
Chr.14: 76151885 - 76214473 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006720191.2 XP_006720254.1 10 - 10 13312 69
XM_017021427.1 XP_016876916.1 11 - 11 13554 69
XM_017021428.1 XP_016876917.1 11 - 11 13539 69


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA C15821.1 - 1 104 Chr. 13 - 39602893 - 39603143 [-] on Build GRCh38 69
GenBankmRNA CB148894.1 - 1 483 Chr. 14 - 76213157 - 76213791 [+] on Build GRCh38 69
GenBankmRNA H94131.1 - 1 121 Chr. 14 - 76213519 - 76213825 [+] on Build GRCh38 69
GenBankmRNA D80932.1 - 1 104 Chr. 14 - 76213536 - 76213783 [+] on Build GRCh38 69
GenBankmRNA DR759654.1 - 1 633 Chr. 14 - 76213007 - 76213870 [+] on Build GRCh38 69
GenBankmRNA C14758.1 - 1 104 Chr. 5 - 168967582 - 168967808 [-] on Build GRCh38 69
GenBankmRNA BF968187.1 - 124 Chr. - - [-] on Build 69
GenBankmRNA AA463371.1 - 1 401 Chr. 14 - 76213239 - 76213715 [+] on Build GRCh38 69
GenBankmRNA R06703.1 - 1 114 Chr. 14 - 76213526 - 76213855 [+] on Build GRCh38 69
GenBankmRNA BX119224.1 - 1 558 Chr. 14 - 76213082 - 76213857 [+] on Build GRCh38 69
GenBankmRNA H62982.1 - 1 148 Chr. 14 - 76213492 - 76213804 [+] on Build GRCh38 69
GenBankmRNA HY282297.1 - 1 Chr. 14 - 76213354 - 76213823 [-] on Build GRCh38 69
GenBankmRNA CR594753.1 - 1 652 Chr. 14 - 76212988 - 76214343 [+] on Build GRCh38 69


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream slit guidance ligand 3 SLIT3 MEGF5, SLIL2, SLIT1, Slit-3, slit2 Sense Chr. - 168661733 - 169301129 on Build GRCh38
Downstream estrogen related receptor beta ESRRB DFNB35, ERR2, ERRb, ESRL2, NR3B2 157.0kb Sense Chr. - 76371347 - 76501839 on Build GRCh38

View Full Product Details