Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK091962.1 - 1 2150 Chr. 3 - 52533968 - 52536252 [+] on Build GRCh38 106
GenBankmRNA AI681702.1 - 1 Chr. 3 - 52535771 - 52536248 [-] on Build GRCh38 106
GenBankmRNA DB320984.1 - 1 Chr. 3 - 52535707 - 52536252 [-] on Build GRCh38 107


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream small integral membrane protein 4 SMIM4 C3orf78 Sense Chr. - 52535986 - 52540570 on Build GRCh38
Downstream polybromo 1 PBRM1 BAF180, PB1 9.1kb Anti-sense Chr. - 52545352 - 52685933 on Build GRCh38

View Full Product Details