The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271893.3 | NP_001258822.1 | 2 - 2 | 87 | |
GenBank mRNA | BC017907.1 | - | 2 - 2 | 87 | |
BC103755.1 | - | 2 - 2 | 87 | ||
DB359681.1 | - | 1 - 1 | 208 | 87 |