Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBXA2R
Entrez Gene ID
6915
Gene Name
thromboxane A2 receptor
Gene Aliases
BDPLT13, TXA2-R
Chromosome Location
Chr.19: 3594506 - 3608749 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001060.5 NP_001051.1 3 - 3 1353 93
XM_011528214.2 XP_011526516.1 4 - 4 3387 93
GenBank mRNA AY429110.1 - 2 - 2 966 93
BC074749.2 - 2 - 2 1005 93
BC074750.2 - 2 - 2 1005 93
BC121148.1 - 2 - 2 611 93
U27325.1 - 3 - 3 1089 93

View Full Product Details